Skip to Content
Merck
All Photos(1)

Documents

32354

Supelco

Kasugamycin hydrochloride

VETRANAL®, analytical standard

Synonym(s):

3-O-[2-Amino-4-[(carboxyiminomethyl)amino]-2,3,4,6-tetradeoxy-D-arabino-hexopyranosyl]-D-chiro-inositol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25N3O9 · HCl
CAS Number:
Molecular Weight:
415.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C14H25N3O9.ClH/c1-3-5(17-12(16)13(23)24)2-4(15)14(25-3)26-11-9(21)7(19)6(18)8(20)10(11)22;/h3-11,14,18-22H,2,15H2,1H3,(H2,16,17)(H,23,24);1H/t3-,4+,5+,6-,7+,8+,9-,10+,11+,14-;/m1./s1

InChI key

ZDRBJJNXJOSCLR-NZXABURVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside
Kasugamycin hydrochloride is an aminoglycoside antibiotic used in the inhibition of both Gram-positive and Gram-negative bacteria. It inhibits protein biosynthesis and prevents the incorporation of amino acids into the protein. It is used as a crop protection agent in controlling rice blast, grey mold, leaf spot and black rot.

Application

Kasugamycin hydrochloride may be used as a reference standard in the determination of kasugamycin hydrochloride in commercial formulations using high-performance capillary electrophoretic coupled with an ultraviolet-visible light detector (HPCE-UV/Vis).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
High-performance capillary electrophoretic method for the determination of antibiotic fungicide kasugamycin in formulated products.
Lo C-C and Hsiao M-Y
Journal of Agricultural and Food Chemistry, 44(8), 2231-2234 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service