Skip to Content
Merck
All Photos(1)

Key Documents

32279

Supelco

C12-C14-Alkyl(ethylbenzyl)dimethylammonium chloride

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$83.60

$83.60


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$83.60

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

$83.60


Estimated to ship on16 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

C12-C14- alkyl(ethylbenzyl)dimethylammonium chloride may be used as a test compound in investigating the cytotoxicity it poses individually or in form of mixtures and the eco-toxicological impact it has on the marine life.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Christen et al.
The Science of the total environment, 586, 1204-1218 (2017-02-27)
Frequently used biocidal disinfectants, including quaternary ammonium compounds (QAC), glutaraldehyde and poly(hexamethylene biguanide) hydrochloride (PHMB), occur in the aquatic environment but their potential effects in fish are poorly known, in particular when occurring as mixtures. To investigate their joint activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service