32066
Amicarthiazol
PESTANAL®, analytical standard
Synonym(s):
2-Amino-4-methyl-1,3-thiazole-5-carboxanilide, 2-Amino-4-methyl-N-phenyl-5-thiazolecarboxamide
About This Item
grade
analytical standard
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
Cc1nc(N)sc1C(=O)Nc2ccccc2
InChI
1S/C11H11N3OS/c1-7-9(16-11(12)13-7)10(15)14-8-5-3-2-4-6-8/h2-6H,1H3,(H2,12,13)(H,14,15)
InChI key
CACOMUHPQMDEJQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service