Skip to Content
Merck
All Photos(2)

Key Documents

32063

Supelco

Dimethylvinphos

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-1-(2,4-dichlorophenyl)vinyl dimethyl phosphate, Dimethylvinfos

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$258.00

$258.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$258.00

About This Item

Empirical Formula (Hill Notation):
C10H10Cl3O4P
CAS Number:
Molecular Weight:
331.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$258.00


Estimated to ship on23 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

COP(=O)(OC)O\C(=C/Cl)c1ccc(Cl)cc1Cl

InChI

1S/C10H10Cl3O4P/c1-15-18(14,16-2)17-10(6-11)8-4-3-7(12)5-9(8)13/h3-6H,1-2H3/b10-6-

InChI key

QSGNQELHULIMSJ-POHAHGRESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Dimethylvinphos may be used as a pesticide reference standard for the determination of the analyte:
  • In edible fungus using gas purge microsyringe extraction (GP-MSE), coupled with on-line gas chromatography-mass spectrometry (GP-MSE-GC-MS).[1]
  • In rice paddies by a method based on the Quick Easy Cheap Effective Rugged and Safe (QuEChERS) sample preparation and gas chromatography with mass spectrometric detection in the selected ion monitoring mode (GC–MS-SIM).[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multi-residue method for the determination of 203 pesticides in rice paddies using gas chromatography/mass spectrometry.
Nguyen TD, et al.
Analytica Chimica Acta, 619(1), 67-74 (2008)
Jingxi Nan et al.
Talanta, 142, 64-71 (2015-05-25)
In this work a new analytical method for a rapid and simultaneous determination of 28 organophosphorus pesticides (OPPs) residues in edible fungus using gas purge microsyringe extraction (GP-MSE), coupled with on-line gas chromatography-mass spectrometry (GP-MSE-GC-MS) has been developed and optimized.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service