Skip to Content
Merck
All Photos(1)

Key Documents

31736

Supelco

Sulfadoxin

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide, Sulfadoxine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$92.10

$92.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$92.10

About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
625453
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$92.10


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC

InChI

1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

PJSFRIWCGOHTNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfadoxin belongs to the long acting group of sulfonamides, which acts by inhibiting dihydropteroate synthase in intracellular sporozoa such as Eimeria and Toxoplasma and Plasmodium.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfadoxin may be used as a reference standard for the determination of the analyte in plasma samples using packed-column supercritical fluid chromatography (SFC).[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Encyclopedic Reference of Parasitology, 2(2) (2001)
Determination of sulfadoxine in human blood plasma using packed-column supercritical fluid chromatography.
Bhoir IS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 757(1) (2001)
Alexia Daoust et al.
NeuroImage, 96, 133-142 (2014-04-08)
The MAP6 (microtubule-associated protein 6) KO mouse is a microtubule-deficient model of schizophrenia that exhibits severe behavioral disorders that are associated with synaptic plasticity anomalies. These defects are alleviated not only by neuroleptics, which are the gold standard molecules for
Feiko O ter Kuile et al.
JAMA, 297(23), 2603-2616 (2007-06-21)
In malaria-endemic regions, strategies to control malaria during pregnancy rely on case management of malaria illness and anemia, and preventive measures such as insecticide-treated nets and intermittent preventive therapy (IPT). To determine the effect of increasing resistance to sulfadoxine-pyrimethamine on
Shane A Pawluk et al.
Clinical pharmacokinetics, 52(3), 153-167 (2013-01-05)
Malaria is one of the most common parasitic infections worldwide. Plasmodium falciparum is the most prevalent strain in Africa and also the most fatal. The disease especially affects children, with those under age 5 years accounting for approximately 86 %

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service