Skip to Content
Merck
All Photos(2)

Key Documents

31189

Supelco

2-Aminobenzimidazole

PESTANAL®, analytical standard

Synonym(s):

2-Benzimidazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
116525
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

226-230 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc2ccccc2[nH]1

InChI

1S/C7H7N3/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H3,8,9,10)

InChI key

JWYUFVNJZUSCSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Zhong et al.
Bioorganic & medicinal chemistry letters, 19(17), 5158-5161 (2009-08-04)
This Letter describes the discovery and key structure-activity relationship (SAR) of a series of 2-aminobenzimidazoles as potent Aurora kinase inhibitors. 2-Aminobenzimidazole serves as a bioisostere of the biaryl urea residue of SNS-314 (1c), which is a potent Aurora kinase inhibitor
Zinc fingered: new compounds that thwart gram-positive biofilm formation by sequestering zinc.
Dinty J Musk
Chembiochem : a European journal of chemical biology, 11(6), 758-760 (2010-03-02)
2-Aminobenzimidazole derivatives strongly inhibit and disperse Pseudomonas aeruginosa biofilms.
Reto Frei et al.
Angewandte Chemie (International ed. in English), 51(21), 5226-5229 (2012-04-11)
S Sudha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 84(1), 184-195 (2011-10-11)
In the present work, we reported a combined experimental and theoretical study on molecular structure, vibrational spectra and HOMO-LUMO analysis of 2-aminobenzimidazole (2-ABD). The FTIR (400-4000 cm(-1)) and FT-Raman spectra (50-3500 cm(-1)) of 2-ABD were recorded. The molecular geometry, harmonic
Ahmad Shaabani et al.
Combinatorial chemistry & high throughput screening, 9(10), 771-776 (2006-12-16)
Tetraheterocyclic benzimidazo[1,2-a]quinazolin-4(1H)-one and tetrahydro-1,2,4-triazolo[5,1-b]quinazolin-8(4H)-one were synthesized in relatively high yields by the condensation reaction of an aldehyde and a cyclic beta-diketone with 2-aminobenzothiazole, 2-aminobenzimidazole or 3-amino-1,2,4-triazole without using any catalyst under solvent-free conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service