Skip to Content
Merck
All Photos(1)

Key Documents

30308

Sigma-Aldrich

Iodic acid

≥99.5% (iodometric)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HIO3
CAS Number:
Molecular Weight:
175.91
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Assay

≥99.5% (iodometric)

impurities

≤0.0005% heavy metals (as Pb)
≤0.0005% iodine (I)

ign. residue

≤0.01% (as SO4)

anion traces

bromide, chloride (as Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

OI(=O)=O

InChI

1S/HIO3/c2-1(3)4/h(H,2,3,4)

InChI key

ICIWUVCWSCSTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manometric carbon determination.
Van Slyke DD and Folch J.
The Journal of Biological Chemistry, 136(2), 509-541 (1940)
Raman studies of iodic acid and sodium iodate.
Durig JR, et al.
The Journal of Physical Chemistry, 69(11), 3886-3892 (1965)
The rate of the reaction between iodic and hydriodic acids.
Dushman S.
The Journal of Physical Chemistry, 8(7), 453-482 (1904)
Allen E Haddrell et al.
The journal of physical chemistry. A, 116(40), 9941-9953 (2012-09-18)
The most used instrument in single particle hygroscopic analysis over the past thirty years has been the electrodynamic balance (EDB). Two general assumptions are made in hygroscopic studies involving the EDB. First, it is assumed that the net charge on
Barbara Krassowska-Swiebocka et al.
Molecules (Basel, Switzerland), 10(2), 394-400 (2007-11-17)
Benzene, halobenzenes, and a number of more or less deactivated arenes,including nitrobenzene, readily reacted in anhydrous HIO3/AcOH/Ac2O/conc. H2SO4 mixtures to probably give ArIO2 intermediates or other hypervalent species (not isolated). The final reaction mixtures were poured into excess aq. Na2SO3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service