Skip to Content
Merck
All Photos(2)

Key Documents

276235

Sigma-Aldrich

3-Noradamantanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$323.00

$323.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$323.00

About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$323.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder or crystals

mp

105-107 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)C12C[C@H]3C[C@H](C[C@H]1C3)C2

InChI

1S/C10H14O2/c11-9(12)10-4-6-1-7(5-10)3-8(10)2-6/h6-8H,1-5H2,(H,11,12)/t6-,7+,8-,10-

InChI key

RXUUYFUQAGICCD-WZXRXUJPSA-N

Application

Building block for "Anti-Bredt"strained molecules.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang-Tian Zhu et al.
Journal of chromatography. A, 1564, 42-50 (2018-06-11)
Tailor-made chitosan fiber was prepared via hydrothermal treatment to serve as a micro-solid phase extraction (micro-SPE) sorbent for the analysis of petroleum acids (PAs) in crude oils. Chitosan fiber, which is commercial and cheap, has a diameter of about 10 μm
Aldrichimica Acta, 18, 82-82 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service