Skip to Content
Merck
All Photos(1)

Key Documents

239577

Sigma-Aldrich

Acetyl chloride

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCl
CAS Number:
Molecular Weight:
78.50
Beilstein:
605303
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.7 (vs air)

vapor pressure

11.69 psi ( 20 °C)
32.33 psi ( 55 °C)

product line

ReagentPlus®

Assay

≥99%

autoignition temp.

1353 °F

expl. lim.

19 %

refractive index

n20/D 1.389 (lit.)

bp

52 °C (lit.)

mp

−112 °C (lit.)

density

1.104 g/mL at 25 °C (lit.)

SMILES string

CC(Cl)=O

InChI

1S/C2H3ClO/c1-2(3)4/h1H3

InChI key

WETWJCDKMRHUPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Frahm et al.
Analytical chemistry, 79(11), 3989-3995 (2007-05-05)
The wide range of protein concentrations found in biological matrixes presents a formidable analytical challenge in proteomics experiments. It is predicted that low-abundance proteins are the likely clinically relevant targets in disease-based proteomics analyses. To effectively analyze low-abundance proteins by
Jacob Bart et al.
Journal of the American Chemical Society, 131(14), 5014-5015 (2009-03-27)
A microfluidic high-resolution NMR flow probe based on a novel stripline detector chip is demonstrated. This tool is invaluable for the in situ monitoring of reactions performed in microreactors. As an example, the acetylation of benzyl alcohol with acetyl chloride
Yu-Ting Liu et al.
The journal of physical chemistry. A, 114(27), 7275-7283 (2010-06-24)
In one-photon dissociation of gaseous acetyl chloride at 248 nm, time-resolved Fourier-transform infrared emission spectroscopy is used to detect the fragments of HCl, CO, and CH(2) in the presence of Ar or O(2). The high-resolution spectra of HCl and CO
Sara J Henderson et al.
Thrombosis and haemostasis, 113(6), 1278-1288 (2015-03-20)
Zinc circulates free in plasma at a concentration of 0.1-2 µM, but its levels increase locally when it is released from activated platelets. Although zinc influences many processes in haemostasis, its effect on fibrinolysis has not been thoroughly investigated. Using
Silvie Van den Hoecke et al.
BMC genomics, 16, 79-79 (2015-03-12)
Influenza viruses exist as a large group of closely related viral genomes, also called quasispecies. The composition of this influenza viral quasispecies can be determined by an accurate and sensitive sequencing technique and data analysis pipeline. We compared the suitability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service