Skip to Content
Merck
All Photos(2)

Key Documents

23035

Sigma-Aldrich

2-Chloroethylamine hydrochloride

purum, ≥98.0% (AT)

Synonym(s):

2-Aminoethyl chloride hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$105.00

$105.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
$105.00

About This Item

Linear Formula:
ClCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
115.99
Beilstein:
3590304
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$105.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

purum

Quality Level

Assay

≥98.0% (AT)

form

solid

mp

140-150 °C (lit.)
140-150 °C

functional group

amine
chloro

SMILES string

Cl.NCCCl

InChI

1S/C2H6ClN.ClH/c3-1-2-4;/h1-2,4H2;1H

InChI key

ONRREFWJTRBDRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroethylamine hydrochloride can be used in the preparation of novel Schiff base ligands. These ligands, when immobilized on silica gels, form inexpensive materials for the removal of metal ions such as Ni2+ and Co2+ ions from contaminated water.[1]
Other applications:
  • Alginate derivatization via aminoethylation to form aminoethyl alginate.[2]
  • Synthesis of N-(2-chloroethyl)-benzaldimine.[3]
  • Derivatization of starch to form aminoethyl (AE-St) starch derivatives.[4]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of polypropylene based hyperbranched absorbent fibers and the study of their adsorption of CO2
Xu T, et al.
Royal Society of Chemistry Advances, 5(42), 32902-32908 (2015)
Production of L (+) lactic acid by Lactobacillus delbrueckii immobilized in functionalized alginate matrices.
Rao C S, et al.
World Journal of Microbiology & Biotechnology, 24(8), 1411-1415 (2008)
Chemical modification of silica gel with synthesized new Schiff base derivatives and sorption studies of cobalt (II) and nickel (II).
Kursunlu A N, et al.
Applied Surface Science, 255(21), 8798-8803 (2009)
Joachim F R Van Guyse et al.
Polymers, 13(3) (2021-02-04)
Smart or adaptive materials often utilize stimuli-responsive polymers, which undergo a phase transition in response to a given stimulus. So far, various stimuli have been used to enable the modulation of drug release profiles, cell-interactive behavior, and optical and mechanical
Khalil Sakeer et al.
Carbohydrate polymers, 163, 108-117 (2017-03-08)
Different starch derivatives were evaluated as supports for attachment and recovery of macrophages (RAW 264.7 line). Gelatinized starch (G-St), acetate starch (Ac-St), carboxymethyl starch and aminoethyl starch were synthesized and characterized by FTIR, 1H NMR, SEM and static water contact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service