Skip to Content
Merck
All Photos(5)

Documents

229253

Sigma-Aldrich

Carminic acid

≥90% (HPLC)

Synonym(s):

Natural Red 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O13
CAS Number:
Molecular Weight:
492.39
Colour Index Number:
75470
Beilstein:
71651
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥90% (HPLC)

form

powder

optical activity

[α]20/D +3.1°, c = 1 in H2O

color

purple to red-purple
red to very dark red

mp

136 °C

solubility

1 N acetic acid: 1%, clear to hazy, red

λmax

495 nm

ε (extinction coefficient)

≥13000 at 222-228  nm in ethanol at 0.03 g/L
≥18000 at 275-281 nm in ethanol at 0.03 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1c(C(O)=O)c(O)cc2C(=O)c3c(O)c(O)c([C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(O)c3C(=O)c12

InChI

1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)/t7-,14-,19+,20-,21+/m1/s1

InChI key

DGQLVPJVXFOQEV-JNVSTXMASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Carmine is a cochineal product usually furnished as an aluminum and calcium salt of carminic acid. Carminic acid is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin. It is suitable for staining nuclei in histological sections. It is utilized in various staining methods including the procedure for detection of glycogen and mucous substances (Best′s carmine), for nuclear staining, or vital staining.

Application

Carminic acid (CI 75470) is used in Orth′s lithium carmine technique for nuclear staining, Best′s stain for glycogen, and Southgates′s mucicarmine method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gabriel J. Lauro, Jack Francis
Natural Food Colorants: Science and Technology (2000)
J.D. Houghton, G.A.F. Hendry
Natural Food Colorants (2012)
Ana Serrano et al.
Analytical and bioanalytical chemistry, 401(2), 735-743 (2011-06-01)
A new analytical approach based on high-performance liquid chromatography with diode array detector (HPLC-DAD) and multivariate data analysis was applied and assessed for analyzing the red dye extracted from cochineal insects, used in precious historical textiles. The most widely used
Samanan Poowakanjana et al.
Journal of food science, 74(9), C707-C712 (2010-05-25)
Carmine used to color surimi seafood often seeps or bleeds from red-colored meat to white meat when vacuum-packed products go through high-temperature long-time pasteurization. Various physical and chemical treatments were examined to investigate means to inhibit or minimize carmine bleeding
Hajime Nakatani et al.
Bioscience, biotechnology, and biochemistry, 75(3), 550-555 (2011-03-11)
The interaction between mammary epithelial and stromal tissue is considered to be important in breast tissue development. In this study, we developed a transplantation procedure for the mammary stromal fibroblastic cell line (MSF) to examine its life in vivo. First

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service