Skip to Content
Merck
All Photos(1)

Key Documents

21931

Sigma-Aldrich

5(6)-Carboxynaphthofluorescein diacetate N-succinimidyl ester

for fluorescence, ≥95% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H23NO11
Molecular Weight:
657.58
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence

Assay

≥95% (HPCE)

solubility

DMSO: soluble

fluorescence

λex 602 nm; λem 672 nm in 0.1 M Tris pH 9.0 (esterase)

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)Oc1cc2Oc3cc(OC(C)=O)c4ccccc4c3C5(OC(=O)c6cc(ccc56)C(=O)ON7C(=O)CCC7=O)c2c8ccccc18


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service