Skip to Content
Merck
All Photos(1)

Key Documents

21639

Supelco

Octanoic acid

analytical standard

Synonym(s):

Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$237.00

$237.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
$237.00

About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747180
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$237.00


Available to ship on09 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 78 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.428 (lit.)
n20/D 1.428

bp

237 °C (lit.)
237-238 °C

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octanoic acid is a medium chain saturated fatty acid (MCFA). It is a nutrient present in dairy products and specific oils like palm kernel and coconut oils.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Octanoic acid may be used as a reference standard in quantification of the analyte in plasma and brain homogenate by high-performance liquid chromatography with ultraviolet detection.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revisiting the metabolism and physiological functions of caprylic acid (C8: 0) with special focus on ghrelin octanoylation.
Lemarie F, et al.
Biochimie, 120, 40-48 (2016)
HPLC analysis of brain and plasma for octanoic and decanoic acids.
Dean HG, et al.
Clinical Chemistry, 35(9), 1945-1948 (1989)
Romain Harmancey et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(10), 3966-3978 (2013-07-05)
Diabetic patients with acute myocardial infarction are more likely to die than nondiabetic patients. In the present study we examined the effect of insulin resistance on myocardial ischemia tolerance. Hearts of rats, rendered insulin resistant by high-sucrose feeding, were subjected
Hong-Fei Tong et al.
Journal of chromatography. A, 1285, 88-96 (2013-03-12)
Hydrophobic charge-induction chromatography (HCIC) is a novel downstream bioprocessing technology for antibody purification and it has several advantages over traditional purification processes. However, its separation selectivity still needs to be improved. In this work, sodium caprylate (NaCA) was used as
K M Morrill et al.
Journal of dairy science, 95(7), 3987-3996 (2012-06-23)
Our objectives were to evaluate the use of refractometry as a means of estimating immunoglobulin G (IgG) concentration of bovine maternal colostrum (MC) and determine if fractionation of MC using caprylic acid (CA) improved estimates of IgG. Samples (n=85) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service