Skip to Content
Merck
All Photos(2)

Key Documents

201820

Sigma-Aldrich

Acid Red 183

Dye content 30 %

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$75.40

$75.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$75.40

About This Item

Empirical Formula (Hill Notation):
C16H11ClN4Na2O8S2 · xCr
CAS Number:
Molecular Weight:
532.84
Colour Index Number:
18800
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$75.40


Estimated to ship on29 May 2025


Request a Bulk Order

form

powder

composition

Dye content, 30%

λmax

494 nm

ε (extinction coefficient)

2900 at 322 nm in H2O at 0.03 g/L
7500 at 244 nm in H2O at 0.03 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Cc1nn(-c2cccc(c2)S([O-])(=O)=O)c3O[Cr](O)Oc4c(cc(Cl)cc4S([O-])(=O)=O)\N=N\c13

InChI

1S/C16H13ClN4O8S2.Cr.2Na.H2O/c1-8-14(19-18-12-5-9(17)6-13(15(12)22)31(27,28)29)16(23)21(20-8)10-3-2-4-11(7-10)30(24,25)26;;;;/h2-7,22-23H,1H3,(H,24,25,26)(H,27,28,29);;;;1H2/q;;2*+1;/p-5

InChI key

AAKJXBYQACYFJH-UHFFFAOYSA-I

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service