Skip to Content
Merck
All Photos(2)

Key Documents

200190

Sigma-Aldrich

Tetrachrome Stain (MacNeal)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$135.00

$135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$135.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$135.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

λmax

524 nm
640 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Cl-].[Cl-].CN(C)c1ccc2nc3ccc(N)cc3[s+]c2c1.CN(C)c4ccc5nc6ccc(cc6[s+]c5c4)N(C)C.[O-]c7c(Br)cc8c(Oc9c(Br)c([O-])c(Br)cc9C8%10OC(=O)c%11ccccc%10%11)c7Br

InChI

1S/C20H8Br4O5.C16H18N3S.C14H14N3S.2ClH.2Na/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;;;;/h1-6,25-26H;5-10H,1-4H3;3-8H,15H2,1-2H3;2*1H;;/q;2*+1;;;2*+1/p-4

InChI key

QXEIULMZMFFXLM-UHFFFAOYSA-J

General description

Tetrachrome stain along with silver stain is used in MacNeal staining method. Tetrachrome stains nucleus blue and silver stain highlight calcification.[1]

Biochem/physiol Actions

Tetrachrome Stain (MacNeal) has been used to stain mouse blood smears to readily identify several morphologically distinct cell types and lineages.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of calcification inhibitors in the pathogenesis of vascular calcification in chronic kidney disease (CKD).
Moe SM
Kidney International, 67, 2295-2304 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service