Skip to Content
Merck
All Photos(1)

Key Documents

199753

Sigma-Aldrich

Toluidine Red

Dye content 70 %

Synonym(s):

Pigment Red 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O3
CAS Number:
Molecular Weight:
307.30
Colour Index Number:
12120
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 70%

mp

270-272 °C (lit.)

λmax

398 nm
507 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(\N=N\c2c(O)ccc3ccccc23)c(c1)[N+]([O-])=O

InChI

1S/C17H13N3O3/c1-11-6-8-14(15(10-11)20(22)23)18-19-17-13-5-3-2-4-12(13)7-9-16(17)21/h2-10,21H,1H3/b19-18+

InChI key

ZLFVRXUOSPRRKQ-VHEBQXMUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CI Pigment Red 3.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 259-267 (1993-01-01)
Ying Jiang et al.
Sexually transmitted diseases, 38(3), 244-245 (2010-09-28)
We retrospectively analyzed the Venereal Disease Research Laboratory test, Treponema pallidum particle agglutination test, and toluidine red unheated serum test (TRUST) in 41 cases of HIV-negative neurosyphilis and 34 non-neurologic syphilitic patient and found that serum-TRUST titers could be the
V Badireenath Konkimalla et al.
Expert review of anticancer therapy, 7(3), 317-329 (2007-03-07)
Oral squamous cell carcinoma ranks among the top ten most common cancers worldwide. Despite the success in diagnosis and therapy during the past 30 years, oral squamous cell carcinoma still belongs to the tumor types with a very unfavorable prognosis.
C E Parham et al.
Journal of clinical microbiology, 20(3), 434-437 (1984-09-01)
The toluidine red unheated serum test (TRUST) antigen, a macroscopic flocculation test antigen developed by Pettit et al. (J. Clin. Microbiol. 18:1141-1145, 1983) by modifying the color-coded antigen of Kasatiya and Lambert (Appl. Microbiol. 28:317-318, 1974), was compared with the
S A Larsen et al.
Journal of clinical microbiology, 17(2), 341-345 (1983-02-01)
Both the rapid plasma reagin (RPR) 18-mm circle card test and the toluidine red unheated serum test (TRUST) were designed to be more rapid than the Venereal Disease Research Laboratory slide test for the serodiagnosis of syphilis. The RPR card

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service