Skip to Content
Merck
All Photos(1)

Key Documents

17194

Millipore

Park and Sanders Selective Supplement II

for Campylobacter spp., suitable for microbiology

Synonym(s):

Campylobacter selective supplement, Selective supplement for thermotolerant Campylobacter

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$326.00

$326.00


Estimated to ship on31 May 2025


Request more information

Select a Size

Change View
5 VIALS
$326.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

$326.00


Estimated to ship on31 May 2025


Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 vials

color

white

application(s)

environmental
food and beverages
veterinary

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

Park and Sanders Selective Supplement II is a specialized microbiological growth medium additive designed to enhance the selective isolation of thermotolerant Campylobacter species in clinical and non-clinical samples. The supplement is optimized for use in conjunction with base media, augmenting the growth of thermotolerant Campylobacter, while suppressing the proliferation of competing flora.

Application

Park and Sanders Selective Supplement II is an antibiotic supplement recommended for the selective isolation and enumeration of thermotolerant Campylobacter species.

Components

Composition (per vial; sufficient for 1000 mL medium)
Cefoperazone 32 mg; Cycloheximide 100 mg

Storage and Stability

Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service