Skip to Content
Merck
All Photos(1)

Key Documents

17170

Millipore

AEA Sporulation Broth (Base), modified

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Product 17170 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ammonium acetate, 1.5 g/L
biopeptone, 10 g/L
disodium phosphate, 4.36 g/L
magnesium sulfate.7H2O, 0.2 g/L
monopotassium phosphate, 0.25 g/L
yeast extract, 10 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.8±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective for Clostridium spp.

Application

For early sporulation of Clostridium perfringens from foods.

Preparation Note

Suspend 26.31 g in 1 litre distilled water. Dispense the medium in 15 ml amounts in screw capped tubes and sterilize by autoclaving at 121°C for 15 minutes. Cool to 45-50°C and add 0.6 ml of filter sterilized 10% raffinose and 0.2 ml each of sterile 0.66 M sodium carbonate and 0.32% cobalt chloride dropwise to each 15 ml base medium in the tubes. Just before use, steam the medium for 10 minutes and after cooling, add 0.2 ml of filter sterilized (freshly prepared) 1.5% sodium ascorbate to each tube of the medium.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joyshree Ganguly et al.
Anaerobe, 63, 102208-102208 (2020-05-11)
Single-cell analysis of microbial population heterogeneity is a fast growing research area in microbiology due to its potential to identify and quantify the impact of subpopulations on microbial performance in, for example, industrial biotechnology, environmental biology, and pathogenesis. Although several

Articles

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

Clostridium perfringens can cause contamination in undercooked or improperly sterilized canned foods and water. Learn to detect, identify, and differentiate this pathogen.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service