Skip to Content
Merck
All Photos(1)

Key Documents

17119

Millipore

Aeromonas Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$138.00

$138.00


Estimated to ship on20 April 2025


Request more information

Select a Size

Change View
5 VIALS
$138.00

About This Item

EC Number:
UNSPSC Code:
41171614
NACRES:
NA.74

$138.00


Estimated to ship on20 April 2025


Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Aeromonas spp.

Application

An antibiotic supplement for the selective isolation of Aeromonas species.

Components

Composition (per vial; sufficient for 500 mL medium)
Ampicillin 2.5 mg. Balanced with carbohydrates/buffer salts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service