Skip to Content
Merck
All Photos(1)

Documents

16316

Sigma-Aldrich

Turkey red oil sodium salt

for microscopy

Synonym(s):

Castor-oil sulfated sodium salt, Sulfated castor oil, Sulforicinolate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
MA.02

grade

for microscopy

Quality Level

description

anionic

Assay

100%

impurities

16-20% water (Karl Fischer)

pH

7-8 (10%)

Looking for similar products? Visit Product Comparison Guide

Other Notes

During long periods of cold weather, and during storage at low temperatures, cloudiness or crystallization can occur in individual cases. These can be eliminated by stirring and gentle heating up to 40°C without any loss of quality.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service