Skip to Content
Merck
All Photos(1)

Key Documents

155012

Sigma-Aldrich

2,2,4-Trimethylpentane

ACS spectrophotometric grade, ≥99%

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12352001
eCl@ss:
39010116
PubChem Substance ID:
grade:
ACS spectrophotometric grade
Assay:
≥99%
technique(s):
UV/Vis spectroscopy: suitable
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)

grade

ACS spectrophotometric grade

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

≥99%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0003 meq/g Water-soluble titr. acid
≤0.005% S compounds

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 220 nm Amax: 0.20
λ: 230 nm Amax: 0.10
λ: 240 nm Amax: 0.04
λ: 250-400 nm Amax: 0.01

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Dolors Benaiges et al.
Biotechnology progress, 26(5), 1252-1258 (2010-10-15)
The characterization of the recombinant Candida rugosa Lip2 (r-Lip2) isoenzyme obtained from fed-batch cultures of Pichia pastoris under PAOX promoter was carried out, determining the optimal pH and temperature as well as their catalytic performance in both hydrolysis and synthesis
Luyan Xue et al.
Colloids and surfaces. B, Biointerfaces, 105, 81-86 (2013-01-29)
Choline acetate is an ionic liquid composed of a kosmotropic anion and a chaotropic cation. According to Hofmeister series, a kosmotropic anion and/or a chaotropic cation could stabilize an enzyme, thereby facilitating the retention of the catalytic activity of the
Nibedita Pal et al.
Analytical chemistry, 83(20), 7736-7744 (2011-09-09)
Fluorescence correlation spectroscopy (FCS) is an ideal tool for measuring molecular diffusion and size under extremely dilute conditions. However, the power of FCS has not been utilized to its best to measure diffusion and size parameters of complex chemical systems.
Yufeng Zhang et al.
Journal of chromatography. A, 1252, 67-73 (2012-07-10)
In this study, a polyethylene Pasteur pipette-based ultrasound-assisted emulsification microextraction (USAEME) applying low-density organic solvent was successfully developed for the extraction of trace levels of organochlorine pesticides (OCPs) in water samples and followed by gas chromatography-mass spectrometry (GC-MS) analysis. In
Kyung Min Park et al.
Food chemistry, 138(2-3), 733-738 (2013-02-16)
In this study, we investigated the possibility of reversed micelles, widely used as an enzyme reactor for lipases, for the determination of lipoxygenase activity. Although it is rapid and simple, reversed micelles have some limitations, such as interference by UV-absorbing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service