Skip to Content
Merck
All Photos(1)

Key Documents

154911

Sigma-Aldrich

2-Methylbutane

spectrophotometric grade, ≥99%

Synonym(s):

Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
spectrophotometric grade
bp:
30 °C (lit.)
vapor pressure:
11.17 psi ( 20 °C)

grade

spectrophotometric grade

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

788 °F

expl. lim.

8.3 %

impurities

<0.020% water

evapn. residue

<0.0003%

refractive index

n20/D 1.354 (lit.)

bp

30 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 192 nm Amax: 1.00
λ: 210 nm Amax: 0.30
λ: 220 nm Amax: 0.07
λ: 240-400 nm Amax: 0.01

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Krustrup et al.
British journal of sports medicine, 43(11), 825-831 (2008-12-23)
To examine the effects of regular participation in recreational soccer on health profile, 36 healthy untrained Danish men aged 20-43 years were randomised into a soccer group (SO; n = 13), a running group (RU; n = 12) and a
Benedikt Galliker et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(25), 6161-6168 (2009-05-14)
We have identified two intermediates in the autoxidation of NO*: ONOO*, which was detected by EPR spectroscopy at 295 K and atmospheric pressure in the gas phase, and ONOONO, a red substance produced at 113 K in 2-methylbutane. The red
Craig M Bullock et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(8), 2188-2200 (2014-04-11)
To investigate the role of the sensory neuropeptide calcitonin gene-related peptide (CGRP) in peripheral sensitization in experimental models of osteoarthritis (OA) pain. Experimental knee OA was induced in rats by intraarticular injection of monosodium iodoacetate (MIA) or by transection of
Nahid A Khan et al.
EMBO molecular medicine, 6(6), 721-731 (2014-04-09)
Nutrient availability is the major regulator of life and reproduction, and a complex cellular signaling network has evolved to adapt organisms to fasting. These sensor pathways monitor cellular energy metabolism, especially mitochondrial ATP production and NAD(+)/NADH ratio, as major signals
Ryo Ogawa et al.
Development (Cambridge, England), 142(1), 51-61 (2014-12-07)
Muscle satellite cells are indispensable for muscle regeneration, but the functional diversity of their daughter cells is unknown. Here, we show that many Pax7(+)MyoD(-) cells locate both beneath and outside the basal lamina during myofiber maturation. A large majority of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service