Skip to Content
Merck
All Photos(1)

Documents

15263

Supelco

Bis(triphenylphosphoranylidene)ammonium chloride

Selectophore

Synonym(s):

Bis(triphenylphosphine)iminium chloride, Hexaphenyldiphosphazenium chloride, PPNCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P=N(Cl)=P(C6H5)3
CAS Number:
Molecular Weight:
574.03
Beilstein:
4097979
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

mp

270-272 °C (lit.)
274-276 °C

storage temp.

2-8°C

SMILES string

ClP(N=P(c1ccccc1)(c2ccccc2)c3ccccc3)(c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/C36H30ClNP2/c37-40(34-25-13-4-14-26-34,35-27-15-5-16-28-35,36-29-17-6-18-30-36)38-39(31-19-7-1-8-20-31,32-21-9-2-10-22-32)33-23-11-3-12-24-33/h1-30H

InChI key

QMTYWEYUVVWPQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Bis(triphenylphosphoranylidene)ammonium chloride was used in two-phase titration and electrochemistry, done for physiochemical characterization of sildenafil. It was also used in cyclic voltammetry reactions, used to study ion transfer (IT) and electron transfer (ET), at a liquid-liquid interface supported on a metallic electrode.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Werner et al.
Talanta, 36(9), 966-968 (1989-09-01)
A nitrate-sensitive electrode based on the ionic associates between bis(triphenylphosphine)iminium and nitrate ions in nitrobenzene solution is proposed. The electrode can be used for determinations of 10(-5) 0.1M nitrate in aqueous solutions.
R Aubeck et al.
The Analyst, 116(8), 811-814 (1991-08-01)
The properties of an ion-selective electrode for indomethacin based on the bis(triphenylphosphoranylidene)ammonium-indomethacin ion-pair complex in a poly(vinyl chloride) membrane are described. The detection limit found for indomethacin at pH 7.0 was 2 x 10(-5) mol l-1 (6.4 micrograms ml-1) and
Physicochemical characterization of sildenafil: ionization, lipophilicity behavior, and ionic-partition diagram studied by two-phase titration and electrochemistry.
Giraulta HH
Helvetica Chimica Acta, 83, 1465-1465 (2000)
Voltammetric determination of extreme standard Gibbs ion transfer energy.
Olaya AJ
Journal of Electroanalytical Chemistry, 644 (1), 60-66 (2010)
Carine Robert et al.
Nature communications, 2, 586-586 (2011-12-14)
The vast majority of commodity materials are obtained from petrochemical feedstocks. These resources will plausibly be depleted within the next 100 years, and the peak in global oil production is estimated to occur within the next few decades. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service