Skip to Content
Merck
All Photos(1)

Key Documents

15180

Supelco

hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate

Selectophore, for ion-selective electrodes, ≥97.0%

Synonym(s):

Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$2,870.00

$2,870.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 G
$2,870.00

About This Item

Empirical Formula (Hill Notation):
C56H84CaO8P2
CAS Number:
Molecular Weight:
987.29
Beilstein:
7405516
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$2,870.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥97.0% (HPLC)
≥97.0%

solubility

THF: 2 mg/10 mL, clear, colorless

SMILES string

C\C(C)=C/CC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)C(C)(C)[C@@H]5CC[C@@]34C
[Ca++].CC(C)(C)CC(C)(C)c1ccc(OP([O-])(=O)Oc2ccc(cc2)C(C)(C)CC(C)(C)C)cc1.CC(C)(C)CC(C)(C)c3ccc(OP([O-])(=O)Oc4ccc(cc4)C(C)(C)CC(C)(C)C)cc3

InChI

1S/2C28H43O4P.Ca/c2*1-25(2,3)19-27(7,8)21-11-15-23(16-12-21)31-33(29,30)32-24-17-13-22(14-18-24)28(9,10)20-26(4,5)6;/h2*11-18H,19-20H2,1-10H3,(H,29,30);/q;;+2/p-2

InChI key

BSGUAHACNRXMLC-UHFFFAOYSA-L

General description

Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt (hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate) is an ionophore of calcium.[1]
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt (hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate) was used for the preparation of ion-selective electrode (ISE) membranes, for an experimental study aimed towards designing, constructing and applying electronic tongue (e-tongue) which operates as a virtual instrument and was based on the array of potentiometric sensors employing the Sequential Injection Analysis technique (SIA).[1] It was also used in Raman and UV-vis spectroscopy, used to determine amounts of a bulky lipophilic cationic additive, tridodecylmethylammonium chloride (TDMACl), present in polyaniline (PANI) membranes.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Calcium-ion sensor in ion-selective electrodes[3][4][5]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.J. Moody et al.
Ion-Selective Electrode Rev., 1, 3-3 (1979)
Virtual instrument for an automated potentiometric e-tongue employing the SIA technique
Duran A
Sensors, 6 (1), 19-29 (2005)
A.J. Frend et al.
Analyst, 108, 1072-1072 (1983)
Raman and UV-vis spectroscopic study of polyaniline membranes containing a bulky cationic additive.
Lindfors T
Journal of Electroanalytical Chemistry, 518 (2), 131-138 (2002)
R W Morton et al.
Analytical biochemistry, 157(2), 345-352 (1986-09-01)
Sensitivity of calcium-selective electrodes heretofore has been limited to calcium concentrations above 10(-8) M in the absence of competing ions. We describe the use of calcium buffers to stabilize the free calcium in the reference electrode. Electrode calibration is linear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service