Skip to Content
Merck
All Photos(1)

Key Documents

14755

Supelco

Bis(dimethylamino)dimethylsilane

for GC derivatization, LiChropur, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]2Si(CH3)2
CAS Number:
Molecular Weight:
146.31
Beilstein:
1736095
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

vapor density

>1 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥95.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.417 (lit.)
n20/D 1.418

bp

128-129 °C (lit.)

mp

−98 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CN(C)[Si](C)(C)N(C)C

InChI

1S/C6H18N2Si/c1-7(2)9(5,6)8(3)4/h1-6H3

InChI key

QULMGWCCKILBTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(dimethylamino)-dimethyl silane was used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterising human electrogenic Na+-cotransporter isoform (hhNBC).[1] It was also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes.[2]

Other Notes

Derivatization reagent for steroids: the (dimethylamino)dimethylsilyl derivatives can be analyzed by GC using a nitrogen-phosphorus detector[3]; Protection of diols, diamines, etc.[4][5]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and characterization of a human electrogenic Na+-cotransporter isoform (hhNBC).
American Journal of Physiology. Cell Physiology, 276 (3), C576-C584 (1999)
Journal of Organometallic Chemistry, 294, 7-7 (1985)
Carbon dioxide transport through membranes.
Missner A
The Journal of Biological Chemistry, 283 (37), 25340-25347 (2008)
R.H. Cragg et al.
Journal of Organometallic Chemistry, 289, 23-23 (1985)
C.F. Poole et al.
Journal of Chromatographic Science, 17, 115-115 (1979)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service