Skip to Content
Merck
All Photos(1)

Key Documents

14135

Supelco

Chlorantraniliprole-(N-methyl-d3)

analytical standard

Synonym(s):

3-Bromo-4′-chloro-1-(3-chloro-2-pyridyl)-2′-methyl-6′-(methyl-d3-carbamoyl)pyrazole-5-carboxanilide, 3-Bromo-N-[4-chloro-2-methyl-6-[(methyl-d3-amino)carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-1H-pyrazole-5-carboxamide, Chlorantraniliprole-d3

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$307.00

$307.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$307.00

About This Item

Empirical Formula (Hill Notation):
C18D3H11BrCl2N5O2
CAS Number:
Molecular Weight:
486.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$307.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

220-240 °C

format

neat

storage temp.

2-8°C

SMILES string

ClC1=CC=CN=C1N2N=C(Br)C=C2C(NC3=C(C(NC([2H])([2H])[2H])=O)C=C(Cl)C=C3C)=O

InChI

1S/C18H14BrCl2N5O2/c1-9-6-10(20)7-11(17(27)22-2)15(9)24-18(28)13-8-14(19)25-26(13)16-12(21)4-3-5-23-16/h3-8H,1-2H3,(H,22,27)(H,24,28)/i2D3

InChI key

PSOVNZZNOMJUBI-BMSJAHLVSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service