All Photos(1)
About This Item
Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
grade
puriss.
Assay
≥97.0%
impurities
≤0.002% heavy metals (as Pb)
≤0.1% total nitrogen (N)
density
3.7 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg
cation traces
Ba: ≤1%
Ca: ≤700 mg/kg
Fe: ≤20 mg/kg
SMILES string
[Sr++].[O-]C([O-])=O
InChI
1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
InChI key
LEDMRZGFZIAGGB-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sunkyung Choi et al.
Environmental science & technology, 40(8), 2608-2614 (2006-05-11)
Radioactive strontium (90Sr) is an important constituent of the complex wastes from past nuclear weapons production and has been stored in underground tanks at U.S. DOE sites (e.g., Hanford, WA). Using bulk and microfocused EXAFS spectroscopy, we examined temporal changes
Wen-Yu Qian et al.
International journal of nanomedicine, 7, 5781-5792 (2012-11-28)
Strontium carbonate nanoparticles (SCNs), a novel biodegradable nanosystem for the pH-sensitive release of anticancer drugs, were developed via a facile mixed solvent method aimed at creating smart drug delivery in acidic conditions, particularly in tumor environments. Structural characterization of SCNs
Geoffrey M Bowers et al.
Solid state nuclear magnetic resonance, 29(1-3), 95-103 (2005-09-20)
The only stable NMR-active isotope of strontium, (87)Sr, is a spin-9/2 quadrupolar nucleus that has a low gyromagnetic ratio, a low natural abundance, and a large nuclear electric quadrupole moment. In this work, we utilize the quadrupolar Carr-Purcell-Meiboom-Gill (QCPMG) pulse
Akiko Sakai et al.
Dental materials journal, 31(2), 197-205 (2012-03-27)
Sr-containing carbonate apatite (SrCAp) specimens of varied Sr contents, ranging from 0 to 13.3 mol%, were prepared through a phosphate treatment of set gypsum-and-carbonate mixture at 100°C for 7 days. Effects of Sr content in SrCAp on microstructure, osteoblast-like cell
Zhaofu Fei et al.
Inorganic chemistry, 44(15), 5200-5202 (2005-07-19)
The crystal structure of a strontium-imidazolium polymeric material obtained from the reaction of zwitterionic 1,3-bis(carboxymethyl)imidazolium with SrCO(3) (2:1) in water is reported. The compound crystallized in the orthorhombic space group Pcc2 with a = 14.545(3) A, b = 8.514(5) A
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service