Skip to Content
Merck
All Photos(1)

Key Documents

12674

Supelco

IQ/OQ/PQ Kit for Ion Chromatography

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$527.00

$527.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$527.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$527.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

certified reference material

technique(s)

ion chromatography: suitable

application(s)

environmental

format

single component solution

General description

IQ/OQ/PQ-Kit for the qualification of ion chromatography systems. Suitable for the verification of linearity, repeatability and carry over.
Contains six certified bromide standards (5, 10, 20, 50, 100 und 1000 mg/kg; 100 mL each). The exact contents, traceability and uncertainty are given in a detailed certificate.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Part of the "IQ/OQ Kit for IC" from Metrohm AG, part number 6.5333.000

Application

This kit enables an efficient and reliable qualification of Ion Chromatography instruments. Find a detailed description of its application in this "Tips & Tricks for IC" article.

Features and Benefits

  • Produced under ISO/IEC 17025 and ISO 17034 accreditation
  • Traceable to SI unit of mole through internationally accepted institutions such as NIST or BAM
  • Accompanied by a comprehensive Certificate of Analysis (CoA) (up to four pages) with data on stability, density, homogeneity, the accuracy of concentration, uncertainty, and traceability
  • Gravimetrically prepared using high-quality qNMR-characterized starting materials and qualified precision balances to ensure minimal uncertainty
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Available in an HDPE bottle, sealed in an aluminum bag

Linkage

Visit the IC Portal to learn more

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service