Skip to Content
Merck
All Photos(1)

Key Documents

12094

Supelco

Docosanoic acid tryptamide

analytical standard

Synonym(s):

N-Docosanoyltryptamine, N-[2-(3-Indolyl)ethyl]docosanamide, BAT, Behenic acid tryptamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54N2O
CAS Number:
Molecular Weight:
482.78
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

116-120 °C

application(s)

food and beverages

format

neat

SMILES string

O=C(NCCC1=CNC2=CC=CC=C12)CCCCCCCCCCCCCCCCCCCCC

InChI

1S/C32H54N2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25-32(35)33-27-26-29-28-34-31-24-22-21-23-30(29)31/h21-24,28,34H,2-20,25-27H2,1H3,(H,33,35)

InChI key

DQCSUFSUVJDOBT-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio Ricci et al.
Nature, 549(7673), 488-491 (2017-09-30)
The majority of the accreting supermassive black holes in the Universe are obscured by large columns of gas and dust. The location and evolution of this obscuring material have been the subject of intense research in the past decades, and
Donghwan Kim et al.
International journal of molecular sciences, 20(14) (2019-07-22)
The high abundance of mitochondria and the expression of mitochondrial uncoupling protein 1 (UCP1) confer upon brown adipose tissue (BAT) the unique capacity to convert chemical energy into heat at the expense of ATP synthesis. It was long believed that
Guillermo Sanchez-Delgado et al.
International journal of obesity (2005), 43(8), 1516-1525 (2018-12-07)
Brown adipose tissue (BAT) seems to play a role in bone morphogenesis. A negative association has been reported between BAT and bone mineral density (BMD) in women, but not in men. A panel of experts has recently published a set
Thiago Matos de Araujo et al.
EBioMedicine, 39, 448-460 (2018-12-14)
The Iroquois homeobox 3 (Irx3) gene has been identified as a functional long-range target of obesity-associated variants within the fat mass and obesity-associated protein (FTO) gene. It is highly expressed in the hypothalamus, and both whole-body knockout and hypothalamic restricted
Zhiqiang Bai et al.
PLoS biology, 15(8), e2002176-e2002176 (2017-08-02)
Enhancing brown fat activity and promoting white fat browning are attractive therapeutic strategies for treating obesity and associated metabolic disorders. To provide a comprehensive picture of the gene regulatory network in these processes, we conducted a series of transcriptome studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service