Skip to Content
Merck
All Photos(1)

Documents

10784

Sigma-Aldrich

Antipyrine

tested according to Ph. Eur.

Synonym(s):

Phenazonum, 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Beilstein:
157775
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

crystals and lumps
powder

mp

109-111 °C (lit.)

solubility

H2O: soluble 1 gm in less than 1ml
alcohol: soluble 1 gm in 1.3 ml
chloroform: soluble 1 gm in 1 ml
diethyl ether: soluble 1 gm in 43 ml

application(s)

pharmaceutical (small molecule)

SMILES string

CN1N(C(=O)C=C1C)c2ccccc2

InChI

1S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

VEQOALNAAJBPNY-UHFFFAOYSA-N

Gene Information

human ... PTGS1(5742)

Looking for similar products? Visit Product Comparison Guide

Application

Antipyrine has been used for immunoblotting. It has also been used as an internal reference marker for studying the transport characteristics of platinum-containing drug, cisplatin in the human placenta in vitro.

Biochem/physiol Actions

Antipyrine also referred to as phenazone or phenazonum is an anti-inflammatory agent that possesses analgesic, antipyretic and platelet-inhibitory actions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eyad Al-Saleh et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 21(10), 726-731 (2008-11-18)
Platinum-containing drugs are used extensively in the treatment of various malignancies in humans. Data are scarce on the maternal-fetal transport characteristics in humans of one such widely used drug, cisplatin, and this prompted us to study its transport characteristics in
E Girbal-Neuhauser et al.
Journal of immunology (Baltimore, Md. : 1950), 162(1), 585-594 (1999-01-14)
Antifilaggrin autoantibodies (AFA) are a population of IgG autoantibodies associated to rheumatoid arthritis (RA), which includes the so-called "antikeratin" Abs and antiperinuclear factor. AFA are the most specific serological markers of RA. We previously showed that they recognize human epidermal
Gro Klitgaard Povlsen et al.
BMC neuroscience, 14, 34-34 (2013-03-19)
Upregulation of vasoconstrictor receptors in cerebral arteries, including endothelin B (ETB) and 5-hydroxytryptamine 1B (5-HT(1B)) receptors, has been suggested to contribute to delayed cerebral ischemia, a feared complication after subarachnoid hemorrhage (SAH). This receptor upregulation has been shown to be
Factors influencing antipyrine elimination.
I H Stevenson
British journal of clinical pharmacology, 4(3), 261-265 (1977-06-01)
Antipyrine as a model drug to study hepatic drug-metabolizing capacity.
H E Poulsen et al.
Journal of hepatology, 6(3), 374-382 (1988-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service