Skip to Content
Merck
All Photos(1)

Key Documents

09575

Sigma-Aldrich

4-Amino-6-(trifluoromethyl)benzene-1,3-disulfonamide

technical, ≥96% (HPLC)

Synonym(s):

5-Amino-α,α,α-trifluorotoluene-2,4-disulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8F3N3O4S2
CAS Number:
Molecular Weight:
319.28
Beilstein:
1164542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥96% (HPLC)

ign. residue

≤2%

mp

241-246 °C

SMILES string

Nc1cc(c(cc1S(N)(=O)=O)S(N)(=O)=O)C(F)(F)F

InChI

1S/C7H8F3N3O4S2/c8-7(9,10)3-1-4(11)6(19(13,16)17)2-5(3)18(12,14)15/h1-2H,11H2,(H2,12,14,15)(H2,13,16,17)

InChI key

KRVABEGPNKGLOT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Brørs et al.
Acta pharmacologica et toxicologica, 51(4), 273-277 (1982-10-01)
The inhibitory effect of hydroflumethiazide (HFT) and its metabolite, 2,4-disulfamyl-5-trifluoromethylaniline (DTA) on cyclic AMP phosphodiesterase and the binding of HFT and DTA to carbonic anhydrase was studied in vitro. Significant inhibition of rat kidney low-Km cyclic AMP phosphodiesterase was observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service