09317
p-Arsanilic acid
purum, ≥98.0% (T)
Synonym(s):
4-Aminobenzenearsonic acid, 4-Aminophenylarsonic acid, 4-Arsanilic acid
About This Item
grade
purum
Assay
≥98.0% (T)
mp
≥300 °C (lit.)
SMILES string
Nc1ccc(cc1)[As](O)(O)=O
InChI
1S/C6H8AsNO3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H,8H2,(H2,9,10,11)
InChI key
XKNKHVGWJDPIRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service