Skip to Content
Merck
All Photos(1)

Key Documents

09178

Supelco

α-Hexylcinnamaldehyde

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$86.00

$86.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$86.00

About This Item

Linear Formula:
C6H5CH=C[(CH2)5CH3]CHO
CAS Number:
Molecular Weight:
216.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$86.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

grade

analytical standard

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.545-1.555
n20/D 1.55 (lit.)

bp

174-176 °C/15 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)\C(CCCCCC)=C\c1ccccc1

InChI

1S/C15H20O/c1-2-3-4-6-11-15(13-16)12-14-9-7-5-8-10-14/h5,7-10,12-13H,2-4,6,11H2,1H3/b15-12+

InChI key

GUUHFMWKWLOQMM-NTCAYCPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Hexylcinnamaldehyde is a stable semisynthetic derivative of cinnamaldehyde. It has a characteristic floral scent, which makes it suitable as a commercial fragrance in perfumes, creams, shampoos, etc.[1]

Application

α-Hexylcinnamaldehyde may be used as an analytical standard for the determination of the analyte in cosmetic products by using a combination of solid-phase microextraction and gas chromatography with flame ionization detector[2] and gas chromatography-mass spectrometry.[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Quantitative determination of some volatile suspected allergens in cosmetic creams spread on skin by direct contact sorptive tape extraction?gas chromatography?mass spectrometry.
Sgorbini B, et al.
Journal of Chromatography A, 1217(16), 2599-2605 (2010)
?-Hexylcinnamaldehyde Synergistically Increases Doxorubicin Cytotoxicity Towards Human Cancer Cell Lines.
Di Giacomo S, et al.
Anticancer Research, 36(7), 3347-3351 (2016)
Validation of SPME-GC-FID method for determination of fragrance allergens in selected cosmetic products
Divisova R, et al.
Acta Chromatographica , 27(3), 509-523 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service