Skip to Content
Merck
All Photos(1)

Key Documents

08319

Supelco

Syringaldehyde

analytical standard

Synonym(s):

3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
182.17
Beilstein:
784514
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

192-193 °C/14 mmHg (lit.)

mp

110-113 °C (lit.)
110-113 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(C=O)cc(OC)c1O

InChI

1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3

InChI key

KCDXJAYRVLXPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Syringaldehyde is an aromatic phenolic aldehyde and a degradation product of lignin. It exhibits antioxidant activity and is reported to inhibit prostaglandin synthetase enzyme. The synthetic form of syringaldehyde is commercially used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.[1][2][3]

Application

Syringaldehyde may be used as an analytical reference standard for the determination of the analyte in guacoextracts and pharmaceutical preparations,(1) cognacs and wines,(2) plum brandies,(4) and wheat straw(5) by various chromatography techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phenolic aldehydes in cognacs and wines by capillary electrophoresis: new cognac quality markers
Panosyan AG, et al.
Journal of Analytical Chemistry, 57(4), 356-361 (2002)
A concise review of the natural existance, synthesis, properties, and applications of syringaldehyde
Ibrahim MNM, et al.
BioResources, 7(3), 4377-4399 (2012)
Development and validation of an LC-MS/MS method with a multiple reactions monitoring mode for the quantification of vanillin and syringaldehyde in plum brandies
Tesevic V, et al.
J. Serb. Chem. Soc., 79(12), 1537-1543 (2014)
Development and validation of a sensitive UPLC-MS/MS instrumentation and alkaline nitrobenzene oxidation method for the determination of lignin monomers in wheat straw
Zheng M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1055(12), 178-184 (2017)
Annekathrin Martina Keiler et al.
European journal of nutrition, 53(4), 1073-1081 (2013-10-31)
The Mediterranean diet rich in fruits, vegetables and olive oil has been related to a lower osteoporosis incidence and accordingly to a reduced fracture risk. These observations might be mediated by the active constituents of extra virgin olive oil, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service