Skip to Content
Merck
All Photos(1)

Documents

07955

Sigma-Aldrich

2-(Phenylthio)aniline

≥98.0% (GC)

Synonym(s):

2-Aminophenyl phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NS
CAS Number:
Molecular Weight:
201.29
Beilstein:
2363643
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

color

gray-brown

mp

43 °C (lit.)

SMILES string

Nc1ccccc1Sc2ccccc2

InChI

1S/C12H11NS/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9H,13H2

InChI key

DGBISJKLNVVJGD-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service