Skip to Content
Merck
All Photos(1)

Key Documents

07270

Sigma-Aldrich

Methyl 6-aminohexanoate hydrochloride

≥99.0% (AT)

Synonym(s):

Methyl 6-aminocaproate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$312.00
100 G
$990.00

$312.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$312.00
100 G
$990.00

About This Item

Linear Formula:
NH2(CH2)5COOCH3 · HCl
CAS Number:
Molecular Weight:
181.66
Beilstein:
3687692
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$312.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (AT)

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

117-124 °C

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)CCCCCN

InChI

1S/C7H15NO2.ClH/c1-10-7(9)5-3-2-4-6-8;/h2-6,8H2,1H3;1H

InChI key

YSLDOTFAFZJPOC-UHFFFAOYSA-N

General description

Methyl 6-aminohexanoate hydrochloride also known as Methyl 6-aminocaproate hydrochloride, is commonly used in the solution-phase peptide synthesis.

Application

Methyl 6-aminohexanoate hydrochloride is used to synthesize papain-catalyzed peptides. [1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Papain-catalyzed synthesis of polyglutamate containing a nylon monomer unit
K Yazawa
Polymers, 8, 194-194 (2016)
Petr Chytil et al.
Molecular pharmaceutics, 15(9), 3654-3663 (2018-03-16)
Herein, the biodegradable micelle-forming amphiphilic N-(2-hydroxypropyl) methacrylamide (HPMA)-based polymer conjugates with the anticancer drug doxorubicin (Dox) designed for enhanced tumor accumulation were investigated, and the influence of their stability in the bloodstream on biodistribution, namely, tumor uptake, and in vivo
Tomáš Etrych et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 58, 1-12 (2014-03-19)
In this study, we describe the synthesis, physico-chemical characterisation and results of the in vitro and in vivo evaluation of the biological behaviour of N-(2-hydroxypropyl)methacrylamide-based (HPMA) copolymer conjugates bearing doxorubicin (DOX) partly bound via a pH-sensitive hydrazone and partly via
Martin Studenovský et al.
Anticancer research, 35(2), 753-757 (2015-02-11)
In the present study, we describe the synthesis and physicochemical properties of a novel pH- and thermoresponsive micellar drug delivery system for an anticancer ellipticinium derivative based on the triblock copolymer poly(ethylene oxide)-block-[tert-butylacrylamide-co-6-(N-methacryloylamino)hexanoic acid hydrazide]-block-poly(ethylene oxide). The system was designed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service