Skip to Content
Merck
All Photos(2)

Key Documents

06934

Supelco

Methyl octanoate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Caprylic acid methyl ester, Methyl caprylate, Octanoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$196.00

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$196.00

About This Item

Linear Formula:
CH3(CH2)6COOCH3
CAS Number:
Molecular Weight:
158.24
Beilstein:
1752270
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.24

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

bp

193-194 °C (lit.)

density

0.877 g/mL at 20 °C (lit.)

format

neat

storage temp.

-10 to -25°C

SMILES string

CCCCCCCC(=O)OC

InChI

1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3

InChI key

JGHZJRVDZXSNKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service