Skip to Content
Merck
All Photos(1)

Key Documents

06922

Supelco

Hexacosanoic acid

analytical standard

Synonym(s):

Cerotic acid, Cerotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)24COOH
CAS Number:
Molecular Weight:
396.69
Beilstein:
1799681
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

format

neat

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)

InChI key

XMHIUKTWLZUKEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexacosanoic acid is classified under the very long chain fatty acids (VLCFAs) group of biologically important compounds.

Application

Hexacosanoic acid may be used as a reference standard for the determination of hexacosanoic acid
  • in human serum and plasma samples for the screening of peroxisomal disorders by gas chromatography-mass spectrometry (GC-MS) operating under selected-ion monitoring (SIM) mode
  • in biological samples by GC-MS
  • in microalgal lipids by solid-phase extraction (SPE) followed by GC-MS analysis
  • as a naphthoxyethyl derivative by high performance liquid chromatography (HPLC) coupled with fluorometric detection

Analysis Note

LC-MS: identity conformes

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Gas chromatography/mass spectrometry analysis of very long chain fatty acids, docosahexaenoic acid, phytanic acid and plasmalogen for the screening of peroxisomal disorders.
Takemoto Y, et al.
Brain & Development, 25(7), 481-487 (2003)
A fluorimetric liquid chromatography for highly sensitive analysis of very long chain fatty acids as naphthoxyethyl derivatives.
Lu CY, et al.
Chromatographia, 51(5-6), 315-321 (2000)
Separation of triacylglycerols and free fatty acids in microalgal lipids by solid-phase extraction for separate fatty acid profiling analysis by gas chromatography.
Paik MJ, et al.
Journal of Chromatography A, 1216(31), 5917-5923 (2009)
High sensitivity quantitative lipidomics analysis of fatty acids in biological samples by gas chromatography-mass spectrometry.
Quehenberger O, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1811(11), 648-656 (2011)
Nicol Kruska et al.
Biochimica et biophysica acta, 1852(5), 925-936 (2015-01-15)
X-linked adrenoleukodystrophy (X-ALD) is a severe neurodegenerative disorder resulting from defective ABCD1 transport protein. ABCD1 mediates peroxisomal uptake of free very-long-chain fatty acids (VLCFA) as well as their CoA-esters. Consequently, VLCFA accumulate in patients' plasma and tissues, which is considered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service