Skip to Content
Merck
All Photos(1)

Key Documents

06212

Supelco

Esculin hydrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O9 · xH2O
CAS Number:
Molecular Weight:
340.28 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -40.5±2.5°, c = 5% in pyridine

shelf life

limited shelf life, expiry date on the label

impurities

≤11% water

application(s)

food and beverages

format

neat

SMILES string

O.OC[C@H]1O[C@@H](Oc2cc3C=CC(=O)Oc3cc2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H16O9.H2O/c16-5-10-12(19)13(20)14(21)15(24-10)23-9-3-6-1-2-11(18)22-8(6)4-7(9)17;/h1-4,10,12-17,19-21H,5H2;1H2/t10-,12-,13+,14-,15-;/m1./s1

InChI key

CQYPGSKIFJFVDQ-QWFKVUSTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Esculin hydrate may be used as an analytical standard for:
  • The detection of glucosidase inhibitors by thin-layer chromatography (TLC) bioautographic method.
  • Library screening in the characterization of novel cellulase genes from uncultured microorganisms.
  • Evaluation of mutagenic and antimutagenic activities of oligorutin and oligoesculin.
  • Detecting β-glucosidase activity in the identification of novel cellulase genes from uncultured microorganisms in rabbit cecum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Feng et al.
Applied microbiology and biotechnology, 75(2), 319-328 (2007-01-12)
A metagenomic cosmid library was prepared in Escherichia coli from DNA extracted from the contents of rabbit cecum and screened for cellulase activities. Eleven independent clones expressing cellulase activities (four endo-beta-1,4-glucanases and seven beta-glucosidases) were isolated. Subcloning and sequencing analysis
Mario O Salazar et al.
Phytochemical analysis : PCA, 18(3), 209-212 (2007-05-16)
A new bioautographic assay suitable for the localisation of beta-glucosidase inhibitors present in a complex matrix is described. Enzyme activity was detected using esculin as the substrate to produce esculetin, which reacts with ferric ion to form a brown complex.
Fengchao Wang et al.
Microbiological research, 164(6), 650-657 (2009-02-24)
Four different environmental DNA libraries were prepared from microbial consortium collected from forest soil, dung of elephant, cow rumen and rotted tree. Seven independent clones specifying cellulase activities (five endo-beta-1,4-glucanases and two beta-glucosidases) were isolated and identified. Sequence analysis of
Ghada Ben Rhouma et al.
Food chemistry, 135(3), 1700-1707 (2012-09-08)
Rutin and esculin have been polymerised by laccase. Five fractions with M(w)¯ between 2127.42 and 8331.85g/mol for oligorutins, and between 688.12 and 6973g/mol for oligoesculins, were obtained. Fourier transformed infrared analysis showed that oligorutins were formed through C-C, C-O and
I Diezhandino et al.
Food chemistry, 168, 134-141 (2014-08-31)
The aim of this work was to study the microbiological, physico-chemical and proteolytic changes in Valdeón blue-veined cheese during ripening. Eight replicas of cheese were produced and a total of 48 cheeses were analysed. Lactic acid bacteria, mainly lactococci, were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service