06069
Oleandrin
analytical standard
Synonym(s):
5β,20(22)-Cardenolide-3β,14,16β-triol-3-([2,6-dideoxy-3-O-methyl-α-L-arabinohexopyranosyl]oxy) 16-acetate, Corrigen, Foliandrin, Folinerin, Neriol, Neriolin, Neriostene
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤5.0% water
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
CO[C@H]1C[C@@H](O[C@@H](C)[C@@H]1O)O[C@H]2CC[C@@]3(C)C(CCC4C3CC[C@]5(C)[C@H]([C@H](C[C@]45O)OC(C)=O)C6=CC(=O)OC6)C2
InChI
1S/C32H48O9/c1-17-29(35)24(37-5)14-27(39-17)41-21-8-10-30(3)20(13-21)6-7-23-22(30)9-11-31(4)28(19-12-26(34)38-16-19)25(40-18(2)33)15-32(23,31)36/h12,17,20-25,27-29,35-36H,6-11,13-16H2,1-5H3/t17-,20+,21-,22-,23+,24-,25-,27-,28-,29-,30-,31+,32-/m0/s1
InChI key
JLPDBLFIVFSOCC-XYXFTTADSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service