Skip to Content
Merck
All Photos(2)

Key Documents

05174

Supelco

DL-Menthol

analytical standard

Synonym(s):

Menthol, 2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$101.00

$101.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$101.00

About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
3194263
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$101.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥98.0% (GC)

optical purity

enantiomeric ratio: ~50:50

shelf life

limited shelf life, expiry date on the label

bp

216 °C (lit.)

mp

34-36 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)C1CCC(C)CC1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling-Ying Wu et al.
Scientific reports, 11(1), 20193-20193 (2021-10-14)
This study aimed to explore the effect of pelvic reconstruction surgery on the relation of pelvic organ prolapse (POP) and overactive bladder (OAB) and the impact of preoperative vaginal oestrogen supplement on vaginal tissue. A total of 100 postmenopausal women
Mohammed Mahmood Sarheed et al.
Frontiers in plant science, 11, 546345-546345 (2020-10-13)
Plants can use volatiles for remote suppression of competitors. Mints produce essential oils, which are known to affect the growth of other plants. We used a comparative approach to identify allelopathic compounds from different Mints (genus Mentha, but also including
Xi Hu et al.
Chemosphere, 113, 30-35 (2014-07-30)
The effects of d-menthol on the growth of Microcystis aeruginosa FACHB-905 and microcystin (MCY) concentration were evaluated by batch culture experiments. The algal biomass and the intracellular and extracellular MCY concentrations were evaluated during 5d incubation. After the d-menthol exposure
Andrew Cheyne et al.
American journal of public health, 104(7), e54-e61 (2014-05-17)
The Family Smoking Prevention and Tobacco Control Act exempted menthol from a flavoring additive ban, tasking the Tobacco Products Safety Advisory Committee to advise on the scientific evidence on menthol. To inform future tobacco control efforts, we examined the public
Menthol cigarette use and stroke risk among US smokers: a critical reappraisal.
Brian Rostron
JAMA internal medicine, 174(5), 808-809 (2014-03-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service