Skip to Content
Merck
All Photos(1)

Key Documents

05094

Sigma-Aldrich

Dodecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

PED-diol (n=12)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$372.00
5 G
$668.00

$372.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$372.00
5 G
$668.00

About This Item

Linear Formula:
HO(CH2CH2O)12H
CAS Number:
Molecular Weight:
546.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$372.00


Available to ship on07 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (oligomer purity)

reaction suitability

reagent type: cross-linking reagent

SMILES string

OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C24H50O13/c25-1-3-27-5-7-29-9-11-31-13-15-33-17-19-35-21-23-37-24-22-36-20-18-34-16-14-32-12-10-30-8-6-28-4-2-26/h25-26H,1-24H2

InChI key

WRZXKWFJEFFURH-UHFFFAOYSA-N

General description

Dodecaethylene glycol also known as PED-diol (n=12), is a versatile cross-linking reagent, used in organic synthesis.

Application

Dodecaethylene glycol can be used as a cross-linking reagent in synthesizing water-soluble forms of tocopherol succinate. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monocomponent hexa-and dodecaethylene glycol succinyl-tocopherol esters: Self-assembly structures, cellular uptake and sensitivity to enzyme hydrolysis
BM Folmer
Biochemical Pharmacology, 78, 1464-1474 (2009)
Eric Waidely et al.
Analytical chemistry, 89(17), 9459-9466 (2017-07-21)
Detection of alpha-l-fucosidase has been shown to have relevance in diagnosing hepatocellular carcinoma. Few assays have been developed to measure this enzyme, with most relying on colorimetric techniques involving the enzyme's kinetics. While these assays are facile and quick, the
Jing Neng et al.
Journal of agricultural and food chemistry, 66(22), 5707-5712 (2018-05-08)
A robust immunoassay based on surface-enhanced Raman scattering (SERS) has been developed to simultaneously detect trace quantities of multiple pathogenic antigens from West Nile virus, Rift Valley fever virus, and Yersinia pestis in fetal bovine serum. Antigens were detected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service