Skip to Content
Merck
All Photos(1)

Key Documents

04872

Supelco

cis-4,10,13,16-Docosatetraenoic acid methyl ester solution

standard for GC, 5% in ethanol

Synonym(s):

Fame 22:4n-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38O2
Molecular Weight:
346.55
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
standard for GC

Assay

≥98.5% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

5% in ethanol

application(s)

food and beverages

format

single component solution

functional group

ester

storage temp.

2-8°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCC\C=C/CCC(=O)OC

InChI

1S/C23H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h7-8,10-11,13-14,19-20H,3-6,9,12,15-18,21-22H2,1-2H3/b8-7-,11-10-,14-13-,20-19-

InChI key

JXKCBSKPWHNCFL-MUDRAGMJSA-N

Application

cis-4,10,13,16-Docosatetraenoic acid methyl ester (Fame 22:4n-6) may be used as an analytical standard in the following:
  • For the identification of the analyte in golden algae, primate brain white matter, and mouse liver using gas chromatography coupled to ion trap mass spectrometry. The double bonds are determined using acetonitrile chemical ionization tandem mass spectrometry.
  • Lipid extract from New Zealand green-lipped mussels using gas chromatography-chemical ionization mass spectrometry (GC-CIMS/MS).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Double bond localization in minor homoallylic fatty acid methyl esters using acetonitrile chemical ionization tandem mass spectrometry
Michaud LA, et al.
Analytical Biochemistry, 307(2), 348-360 (2002)
Gas chromatography?chemical ionization?mass spectrometric fatty acid analysis of a commercial supercritical carbon dioxide lipid extract from New Zealand green?lipped mussel (Perna canaliculus)
Wolyniak JC, et al.
Lipids, 40(4), 355-360 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service