Skip to Content
Merck
All Photos(1)

Key Documents

04831

Supelco

Abberior® STAR 470SXP, biotin

≥90.0% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥90.0% (HPCE)

manufacturer/tradename

Abberior GmbH

fluorescence

λex 472 nm; λem 598 nm±10 nm in DMSO

detection method

fluorometric

storage temp.

2-8°C

Suitability

Designed and tested for fluorescent super-resolution microscopy

Legal Information

abberior is a registered trademark of Abberior GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service