Skip to Content
Merck
All Photos(1)

Key Documents

03716

Supelco

(±)-Lisofylline

analytical standard

Synonym(s):

Lisofylline, (±)-1-(5-Hydroxyhexyl)-3,7-dimethylxanthine, 5-Hydroxy pentoxifylline, Penthydroxyfylline

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$254.00

$254.00


Estimated to ship on04 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C13H20N4O3
CAS Number:
Molecular Weight:
280.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Estimated to ship on04 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O=C1N(CCCCC(O)C)C(N(C)C2=C1N(C)C=N2)=O

InChI

1S/C13H20N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8-9,18H,4-7H2,1-3H3

InChI key

NSMXQKNUPPXBRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lisofylline (LSF) is a synthetically modified methylxanthine which is known to exhibit various therapeutic properties. It is a potent anti-inflammatory agent and an antioxidant with the ability to suppress oxygen radical production. LSF is used for treating acute lung injury and also has a role in the early treatment of diabetes.[1][2][3][4]

Application

Lisofylline may be used as an analytical reference standard for the determination of the analyte in rat plasma[5][6] by various high-performance liquid chromatography (HPLC) based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous estimation of lisofylline and pentoxifylline in rat plasma by high performance liquid chromatography-photodiode array detector and its application to pharmacokinetics in rat
Italiya KS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1061(1), 49-56 (2017)
Lisofylline: a potential lead for the treatment of diabetes
Yang Z, et al.
Biochemical Pharmacology, 69(1), 1-5 (2005)
Validation of a high
Walczak M, et al.
Acta Poloniae Pharmaceutica, 66(3), 215-224 (2009)
Combined treatment with lisofylline and exendin-4 reverses autoimmune diabetes
Yang Z, et al.
Biochemical and Biophysical Research Communications, 344(3), 1017-1022 (2006)
Lisofylline, a novel antiinflammatory agent, protects pancreatic $\beta$-cells from proinflammatory cytokine damage by promoting mitochondrial metabolism
Chen M, et al.
Endocrinology, 143(6), 2341-2348 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service