Skip to Content
Merck
All Photos(1)

Key Documents

03674

Sigma-Aldrich

Boc-β-Homoarg(Tos)-OH

≥98.0% (TLC)

Synonym(s):

Nβ-Boc-Nω-tosyl-L-β−homoarginine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$972.00

$972.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$972.00

About This Item

Empirical Formula (Hill Notation):
C19H30N4O6S
CAS Number:
Molecular Weight:
442.53
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$972.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (TLC)

form

crystalline

color

light yellow

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(=O)(=O)NC(=N)NCCC[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

InChI

1S/C19H30N4O6S/c1-13-7-9-15(10-8-13)30(27,28)23-17(20)21-11-5-6-14(12-16(24)25)22-18(26)29-19(2,3)4/h7-10,14H,5-6,11-12H2,1-4H3,(H,22,26)(H,24,25)(H3,20,21,23)/t14-/m0/s1

InChI key

RYHLUZMBVLFVPG-AWEZNQCLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service