Skip to Content
Merck
All Photos(1)

Key Documents

03561

Supelco

Silica gel Inorganic Sorbent

high-purity grade, 90-200 mesh, FIA according to DIN 51791

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$1,280.00

$1,280.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$1,280.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

$1,280.00


Check Cart for Availability

Request a Bulk Order

Product Name

Silica gel, high-purity grade, FIA according to DIN 51791

grade

high-purity grade

form

powder

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

0.071-0.16 mm

pore size

60 Å mean pore size

pH

5.5-7.5

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for the determination of the content of hydrocarbon groups in liquid fuels according to DIN 51791

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service