Skip to Content
Merck
All Photos(1)

Key Documents

03369

Supelco

Nickel atomic spectroscopy standard concentrate 10.00 g Ni

10.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Nickel(II) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: catalyst
core: nickel

packaging

ampule of

concentration

10.00 g/L

solubility

H2O: soluble

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with NiCl2.6H2O and H2O

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeinhom M El-Bahy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 92, 105-112 (2012-03-27)
Cast technique was used to prepare films of sodium carboxymethyl cellulose (CMC) doped with different ratios of NiCl(2)·6H(2)O in the range of 0-40Ni(2+) wt.%. Thermal analysis (DTA) in the range of 25-600°C and dielectric properties in the temperature range of
Siyue Lou et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 51, 251-258 (2012-09-20)
Nickel, a metal commonly found in battery plants and welding factories, has potential cardiotoxicity, while all-trans retinoid acid (atRA) can promote cardiovascular repair and myocardial recovery. The purpose of this study was to investigate whether atRA could prevent cardiotoxicity induced
Terran E Bergdale et al.
Journal of biotechnology, 161(3), 181-189 (2012-07-14)
Microbiologically induced calcium carbonate precipitation (MICCP) is a naturally occurring biological process that has shown its potential in remediation of a wide range of structural damages including concrete cracks. In this study, genetically engineered microorganisms, capable of producing extracellular polymeric
Abhay K Pandey et al.
Neuroscience letters, 517(2), 128-132 (2012-05-09)
Bisphenol-A (BPA), a toxic chemical from polycarbonate plastics, is known for behavioural and neural abnormalities. These neuro-behavioural changes reflect the changes in neural activity. However the effect of BPA on nerve action potential is not available. Therefore, present investigation was
Alireza Pourkhabbaz et al.
Biomedical and environmental sciences : BES, 24(6), 656-660 (2012-03-01)
To determine the effects of water hardness on the toxicities of cobalt (Co) and nickel (Ni) to a freshwater fish, Capoeta fusca. Toxicity was investigated by static bioassay. Fish were exposed to cobalt (as CoCl(2)) and nickel (as NiCl(2)) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service