Skip to Content
Merck
All Photos(1)

Key Documents

02336

Millipore

PALCAM Listeria Selective Supplement according to Van Netten et al.

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$91.30

$91.30


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
5 VIALS
$91.30

About This Item

MDL number:
UNSPSC Code:
41171614
NACRES:
NA.85

$91.30


Estimated to ship on30 May 2025


Request more information

Agency

according to GB 4789.30-2016

Quality Level

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
pathogen testing

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

Looking for similar products? Visit Product Comparison Guide

Application

An antimicrobial supplement recommended for the selective isolation and identification of Listeria monocytogenes.

Components

(per vial, sufficient for 500 mL medium)
Polymyxin B sulfate 50000 IU
Ceftazidime 10.0 mg
Acriflavine hydrochloride 2.5 mg

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service