Skip to Content
Merck
All Photos(1)

Key Documents

01479

Supelco

2-Hydroxy-4-methoxybenzophenone

certified reference material, TraceCERT®

Synonym(s):

Oxybenzone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)COC6H5
CAS Number:
Molecular Weight:
228.24
Beilstein:
1913145
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

150-160 °C/5 mmHg (lit.)

mp

62-64 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COc1ccc(c(O)c1)C(=O)c2ccccc2

InChI

1S/C14H12O3/c1-17-11-7-8-12(13(15)9-11)14(16)10-5-3-2-4-6-10/h2-9,15H,1H3

InChI key

DXGLGDHPHMLXJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K U Schallreuter et al.
The Journal of investigative dermatology, 106(3), 583-586 (1996-03-01)
We used noninvasive Fourier transform (FT) Raman spectroscopy to follow the fate of the broadly used ultraviolet UVA sun blocker, oxybenzone, after topical application to the skin. Our results showed that oxybenzone is rapidly photo-oxidized, yielding oxybenzone semiquinone, a potent
Pablo Gago-Ferrero et al.
The Science of the total environment, 443, 209-217 (2012-11-29)
The goal of this study was to bring forward new data and insights with respect to the effect of operational variables and reaction pathways during ozonation and peroxone oxidation of the UV filter compound benzophenone-3 (BP3) in water. A systematic
Pablo Gago-Ferrero et al.
The Science of the total environment, 427-428, 355-363 (2012-05-01)
Photodecomposition might be regarded as one of the most important abiotic factors affecting the fate of UV absorbing compounds in the environment and photocatalysis has been suggested as an effective method to degrade organic pollutants. However, UV filters transformation appears
Jessica H Savage et al.
The Journal of allergy and clinical immunology, 130(2), 453-460 (2012-06-19)
Endocrine-disrupting compounds (EDCs) have immune-modulating effects. We were interested in determining their association with allergic sensitization. We sought to determine the association between EDCs and allergic sensitization and whether this relationship depends on the antimicrobial properties of the EDCs, sex
Chi-Feng Hung et al.
Journal of dermatological science, 68(3), 135-148 (2012-10-03)
Ultraviolet (UV) exposure is the predominant cause of skin aging. A systematic evaluation of drug skin permeation via photoaged skin is lacking. The aim of this work was to investigate whether UVA and UVB affect absorption by the skin of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service